missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTLL10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26014-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Inactive polyglycylase.Especificaciones
TTLL10 | |
Polyclonal | |
Unconjugated | |
TTLL10 | |
Inactive polyglycylase TTLL10, TTLL10, TTLL5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
254173, 298692, 330010 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A4Q9F3, Q5XI57, Q6ZVT0 | |
TTLL10 | |
TTLL10 Fusion Protein Ag23258 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |