missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18974-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility.Especificaciones
TSPAN13 | |
Polyclonal | |
Unconjugated | |
TSPAN13 | |
NET 6, NET6, Tetraspan NET 6, tetraspanin 13, TM4SF13, Tspan 13, TSPAN13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27075, 366602, 66109 | |
-20°C | |
Liquid |
Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95857, Q5FVL6, Q9D8C2 | |
Tspan13 | |
TSPAN13 Fusion Protein Ag6997 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |