missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptase (Mast Cell Marker) Mouse, Clone: TPSAB1/1961, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 7177-MSM1-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Tryptase (Mast Cell Marker) | |
Secreted and Cytoplasmic | |
TPSAB1/1961 | |
Unconjugated | |
Mouse | |
31-36kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
7177 | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human Tryptase protein fragment (apprx. aa 115-233) | |
Skin or Tonsil | |
20 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |