missing translation for 'onlineSavingsMsg'
Learn More

TROP-2 Antibody (TACSTD2/2151), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-75744B

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215998

  • 578.82€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

TROP-2 Monoclonal specifically detects TROP-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

TROP-2
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
TACSTD2
Recombinant fragment of human TROP-2 protein (around aa 31-274) (exact sequence is proprietary) (Uniprot: P09758 )
0.1 mL
Signal Transduction
TROP-2 is a cell surface glycoprotein receptor. It is a single pass type I membrane protein containing one thyroglobulin type-1 domain, an epidermal growth factor-like repeat, a phosphatidylinositol binding site and tyrosine phosphorylation sites near the C-terminus. It plays a role in transducing intracellular calcium signals. It is expressed in trophoblast cells, cornea and multi-stratified epithelia. It is also highly expressed in several types of tumors and is involved in regulating the growth of carcinoma cells.
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TACSTD2/2151
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2
Mouse
Protein A or G purified
RUO
Primary
Human
IgG2b κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales