Learn More
Tris(triphenylphosphine)rhodium(I) chloride, 99.99%, (trace metal basis), ACROS Organics™
Marca:  Acros Organics 194570010
Detalles adicionales : CAS : 14694-95-2 Peso : 0.05100kg
Cantidad. 1g
Envase Glass bottle
 Ver más versiones de este producto
Código de producto. 10646212
  • 80.30€ / 1 g
Fecha de envío estimada
del proveedor 01-03-2019
Añadir a la cesta
Descripción
Descripción
For ultra trace analysis
Especificación
14694-95-2
925.231
QBERHIJABFXGRZ-UHFFFAOYSA-M
84599
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Rh]
C54H45ClP3Rh-
MFCD00010016
chlorotris triphenylphosphine rhodium i, chlorotris triphenylphosphine rhodium, tris triphenylphosphine rhodium i chloride, tris triphenylphosphine chlororhodium, wilkinson's catalyst, unii-0fv534bkev, tris triphenylphosphine rhodium chloride, 0fv534bkev, tris triphenylphosphine rhodium i chloride, rhodium; triphenylphosphane; chloride
rhodium;triphenylphosphane;chloride
Especificación
Tris(triphenylphosphine)rhodium(I) chloride
Glass bottle
245.0°C to 250.0°C
99.99%
99.99%
[(C6H5)3P]3RhCl
01,140; 02,448; 03,325; 04,559; 05,736; 06,652; 07,68; 08,109; 09,113; 15,90
chlorotris triphenylphosphine rhodium i, chlorotris triphenylphosphine rhodium, tris triphenylphosphine rhodium i chloride, tris triphenylphosphine chlororhodium, wilkinson's catalyst, unii-0fv534bkev, tris triphenylphosphine rhodium chloride, 0fv534bkev, tris triphenylphosphine rhodium i chloride, rhodium; triphenylphosphane; chloride
QBERHIJABFXGRZ-UHFFFAOYSA-M
rhodium;triphenylphosphane;chloride
84599
Crystalline Powder
Trace Metal Basis
Authentic
Maroon
1g
14694-95-2
C54H45ClP3Rh-
MFCD00010016
15, 10245
Solubility in water: insoluble. Other solubilities: soluble in chloroform, dichloromethane, benzene,, toluene, less soluble in acetic acid, ketones, methanol and, lower aliphatic alcohols, virtually insoluble in alkanes and cyclohexane
C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-].[Rh]
925.231
925.23
99.99%
Videos
SDS Documentación