missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2'-bipiridina)diclororutenio (II) hexahidrato, cristalino rojo-anaranjado, >98 %, Thermo Scientific Chemicals

Catalog Number p-4782213
Click to view available options
Cantidad:
0.5 g
2 g
50525-27-4
C30H36Cl2N6O6Ru
748.62
MFCD00149670
WHELTKFSBJNBMQ-UHFFFAOYSA-L
tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate
131664161
λ²-ruthenium(2+) tris(2,2'-bipyridine) hexahydrate dichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1
50525-27-4
C30H36Cl2N6O6Ru
748.62
MFCD00149670
WHELTKFSBJNBMQ-UHFFFAOYSA-L
tris 2,2'-bipyridyl ruthenium ii chloride hexahydrate
131664161
λ²-ruthenium(2+) tris(2,2'-bipyridine) hexahydrate dichloride
O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1

Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate is a catalyst used in light-induced redox reactions. It is an important catalyst in studies towards photochemical water splitting. This product can be used as a catalyst in pharmaceutical research. It is widely applied in fields of photoelectrical chemical components, photovoltaic cells, smart biosensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Tris(2, 2'-bipyridine)dichlororuthenium(II) hexahydrate
Punto de fusión 63°C to 66°C
Color Rojo a naranja
Intervalo de porcentaje de ensayo >98%
Cantidad 0.5 g
Información de solubilidad Soluble in water.
Formula Weight (peso de la fórmula) 748.63 (640.54 Anhydrous)
Forma física cristalino/a/s

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'