missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetrametil-3,5-heptanodionato)bismuto(III), 99,9 %, Thermo Scientific Chemicals

Catalog Number 11309567
Click to view available options
Cantidad:
250 mg
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
142617-53-6
C33H57BiO6
758.793
MFCD00064763
ZNHBPQSSVCRFST-LWTKGLMZSA-K
tris 2,2,6,6-tetramethyl-3,5-heptanedionato bismuth iii, bismuth iii tetramethylheptanedionate, 4z-5-bis 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy bismuthanyl oxy-2,2,6,6-tetramethylhept-4-en-3-one
16717622
(Z)-5-bis[[(Z)-2,2,6,6-tetrametil-5-oxohept-3-en-3-il]oxi]bismutaniloxi-2,2,6,6-tetrametilhept-4-en-3-ona
CC(C)(C)C(=CC(=O)C(C)(C)C)O[Bi](OC(=CC(=O)C(C)(C)C)C(C)(C)C)OC(=CC(=O)C(C)(C)C)C(C)(C)C
Este artículo no se puede devolver. Vea la política de devoluciones
142617-53-6
C33H57BiO6
758.793
MFCD00064763
ZNHBPQSSVCRFST-LWTKGLMZSA-K
tris 2,2,6,6-tetramethyl-3,5-heptanedionato bismuth iii, bismuth iii tetramethylheptanedionate, 4z-5-bis 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy bismuthanyl oxy-2,2,6,6-tetramethylhept-4-en-3-one
16717622
(Z)-5-bis[[(Z)-2,2,6,6-tetrametil-5-oxohept-3-en-3-il]oxi]bismutaniloxi-2,2,6,6-tetrametilhept-4-en-3-ona
CC(C)(C)C(=CC(=O)C(C)(C)C)O[Bi](OC(=CC(=O)C(C)(C)C)C(C)(C)C)OC(=CC(=O)C(C)(C)C)C(C)(C)C

Used as pharamaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)bismuth(III)
Punto de fusión 114°C to 116°C
Color Blanco a marrón
Punto de ebullición 150°C (0.05 mmHg)
Cantidad 250 mg
Sensibilidad Air and moisture sensitive
Información de solubilidad Insoluble in water.
Formula Weight (peso de la fórmula) 758.79
Porcentaje de pureza 99.9%
Forma física cristalino/a/s

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'