Learn More
Thermo Scientific™ Paquete esterilizado de triple envoltura con indicador de peróxido de hidrógeno vaporizado (VHP) y agar dextrosa Sabouraud
Lleve a cabo el control medioambiental en sus entornos más regulados mediante placas irradiadas de triple envoltura Thermo Scientific™, para aplicaciones de muestreo del aire activas y pasivas.
Marca:  Thermo Scientific™ PO5502B
Detalles adicionales : Peso : 2.50000kg
 Ver más versiones de este producto
Código de producto. 15446030
  • 316.00€ / paquete de 100
Fecha estimada de envío
19-01-2021
Añadir a la cesta
Descripción
Descripción
Achieve confidence in your results and ensure optimum media fertility using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.
Three protective layers reduce risk of contamination:
  • Primary film with moisture control patch
  • Secondary film VHP barrier with immobilized desiccant sachet
  • Tertiary film dust cover
The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture
Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs
Not all products are available for sale in all territories. Please inquire.
Spezifikation
Spezifikation
Ivory, transparent
25.0g ± 0.5g
Agar
12 Weeks
90mm Monoplate
Triple Wrap Sterile Pack w/VHP Indicator Sabouraud Dextrose Agar
2° to 25°C
Germany
100/Pk.
Triple Wrapped - Irradiated
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation