missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIP13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19602-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Thyroid hormone receptors (TRs) are transcription factors that regulate the expression of specific genes in a hormone-dependent manner. TRIP13 (thyroid hormone receptor interactor 13), also called 16E1BP, is a transcription factor that interacts with the ligand binding domain of the thyroid receptor (TR) as well as a variety of target genes including human papilloma virus type 16 (HPV16) E1. Unlike most TRIP proteins which function only in the presence of hormones, TRIP13 does not require the presence of thyroid hormone to interact with TR. The association of TRIP13 with (HPV16) E1 suggests that TRIP13 may have tumor suppressor gene function. TRIP13 is a 432 amino acid protein with 2 different isoforms produced by alternative splicing.Especificaciones
TRIP13 | |
Polyclonal | |
Unconjugated | |
TRIP13 | |
16E1 BP, 16E1BP, PCH2, TR interacting protein 13, TRIP 13, TRIP13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
292206, 69716, 9319 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15645, Q3UA06, Q5XHZ9 | |
Trip13 | |
TRIP13 Fusion Protein Ag6595 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |