missing translation for 'onlineSavingsMsg'
Learn More

TRIM Antibody (TRIM-04), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NB500-485UV

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215850

  • 491.35€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

TRIM Monoclonal specifically detects TRIM in Human samples. It is validated for Western Blot, Flow Cytometry.
Especificaciones

Especificaciones

TRIM
Monoclonal
DyLight 350
pp29/30, T cell receptor associated transmembrane adaptor 1, T cell receptor interacting molecule, T-cell receptor interacting molecule, T-cell receptor-associated transmembrane adapter 1, T-cell receptor-interacting molecule, TCRIM, TRIMHSPC062
Mouse
Affinity Purified
RUO
Primary
The antibody TRIM-04 reacts with T cell receptor-interacting molecule (TRIM), a 30 kDa adaptor protein expressed by T cells.
Store at 4C in the dark.
Western Blot, Flow Cytometry
TRIM-04
Western Blot, Flow Cytometry
TRAT1
Recombinant fragment, corresponding to amino acids 29-186 of Human TRIM.
0.1 mL
Adaptive Immunity, Immunology
50852
Human
IgG2a
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only