missing translation for 'onlineSavingsMsg'
Learn More

Óxido de trietilfosfina, Thermo Scientific Chemicals

Catalog Number p-7049860
Click to view available options
Cantidad:
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
597-50-2
C6H15OP
134.16
MFCD00013914
ZSSWXNPRLJLCDU-UHFFFAOYSA-N
triethylphosphine oxide, phosphine oxide, triethyl, triethylphoshorus oxide, c2h5 3p=o, triethylphospine oxide, et3po, triethylphosphino-1-one, triethyl oxo phosphorane, diethylphosphoryl ethane, triethylphosphine oxide 1g
79061
1-dietilfosforiletano
CCP(=O)(CC)CC
Este artículo no se puede devolver. Vea la política de devoluciones
597-50-2
C6H15OP
134.16
MFCD00013914
ZSSWXNPRLJLCDU-UHFFFAOYSA-N
triethylphosphine oxide, phosphine oxide, triethyl, triethylphoshorus oxide, c2h5 3p=o, triethylphospine oxide, et3po, triethylphosphino-1-one, triethyl oxo phosphorane, diethylphosphoryl ethane, triethylphosphine oxide 1g
79061
1-dietilfosforiletano
CCP(=O)(CC)CC

Triethylphosphine oxide is used as a probe molecule and systems are evaluated by 31P NMR spectroscopy. Triethylphosphine oxide is used to produce tris(perfluoroethyl)difluorophosphane. This reaction will need reagent of HF. This reaction needs electrochemical fluorination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Triethylphosphine oxide
Punto de fusión 48°C to 50°C
Color Blanco
Cantidad 5 g
Información de solubilidad Soluble in water.
Formula Weight (peso de la fórmula) 134.16
Forma física cristalino/a/s

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'