missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Óxido de trietilfosfina, Thermo Scientific Chemicals
Click to view available options
Cantidad:
5 g
Descripción
Triethylphosphine oxide is used as a probe molecule and systems are evaluated by 31P NMR spectroscopy. Triethylphosphine oxide is used to produce tris(perfluoroethyl)difluorophosphane. This reaction will need reagent of HF. This reaction needs electrochemical fluorination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Triethylphosphine oxide |
Punto de fusión | 48°C to 50°C |
Color | Blanco |
Cantidad | 5 g |
Información de solubilidad | Soluble in water. |
Formula Weight (peso de la fórmula) | 134.16 |
Forma física | cristalino/a/s |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'