Learn More
Hidrobromuro de trietilamina, 99 %, Thermo Scientific Chemicals
Catalog Number p-204011
Click to view available options
Cantidad:
100 g
500 g
Envase:
Frasco de vidrio
636-70-4
C6H16BrN
182.11
MFCD00054287
NRTLTGGGUQIRRT-UHFFFAOYSA-N
triethylamine hydrobromide, ethanamine, n,n-diethyl-, hydrobromide, ethanamine, n,n-diethyl-, hydrobromide 1:1, n,n-diethylethanamine hydrobromide, acmc-20ajeg, et3n.hbr, triethylammonium bromide salt, triethylamine hydrobromide salt
2734074
[Br-].CC[NH+](CC)CC
636-70-4
C6H16BrN
182.11
MFCD00054287
NRTLTGGGUQIRRT-UHFFFAOYSA-N
triethylamine hydrobromide, ethanamine, n,n-diethyl-, hydrobromide, ethanamine, n,n-diethyl-, hydrobromide 1:1, n,n-diethylethanamine hydrobromide, acmc-20ajeg, et3n.hbr, triethylammonium bromide salt, triethylamine hydrobromide salt
2734074
[Br-].CC[NH+](CC)CC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Triethylamine hydrobromide
Nombre de nota 99%
Color Blanco
Temperatura de inflamación >65°C
Espectro infrarrojo Auténtico
Intervalo de porcentaje de ensayo 98.5% min. (Argentometry)
Envase Frasco de vidrio
Fórmula lineal (C2H5)3N·HBr
Cantidad 100 g
Nombre IUPAC bromuro de trietilazanio
Formula Weight (peso de la fórmula) 182.1
Porcentaje de pureza 99%
Forma física Polvo cristalino o copos
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • Triethylamine hydrobromide
Palabra de advertencia
  • Atención
Clasificación
  • Lesión ocular grave/irritación ocular Categoría 2