missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 4-phosphonocrotonate, cis + trans, 94%

Product Code. 11424443
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
Unit Size:
10g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424443 250 g 250g
11414443 10 g 10g
11433920 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424443 Supplier Thermo Scientific Alfa Aesar Supplier No. A10995.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10236-14-3 | C10H19O5P | 250.23 g/mol

Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl 4-phosphonocrotonate is used as a reactant for iminodipropionic acid as a leaving group for DNA polymerization by HIV-1 reverse transcriptase, orally bioavailable GPR40 agonist synthesis, preparation of mGlu4R agonists and synthesis of fluoroketone inhibitors of group VIA calcium-independent phospholipase A2.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10236-14-3
Molecular Formula C10H19O5P
Molecular Weight (g/mol) 250.23
MDL Number MFCD00009192
InChI Key LXLODBXSCRTXFG-BQYQJAHWSA-N
Synonym triethyl 4-phosphonocrotonate, e-ethyl 4-diethoxyphosphoryl but-2-enoate, ethyl 4-diethoxyphosphoryl but-2-enoate, triethyl .gamma.-phosphonocrotonate, ethyl 4-diethylphosphono crotonate, ethyl 2e-4-diethoxyphosphoryl but-2-enoate, ethyl e-4-diethoxyphosphorylbut-2-enoate, ethyl 4-diethylphosphonocrotonate, ethyl trans-4-diethylphosphono crotonate, 4-diethoxyphosphinyl crotonic acid ethyl ester
PubChem CID 5365783
IUPAC Name ethyl (E)-4-diethoxyphosphorylbut-2-enoate
SMILES CCOC(=O)\C=C\CP(=O)(OCC)OCC

Specifications

Density 1.13
Boiling Point 133°C to 135°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.455
Quantity 250 g
Formula Weight 250.23
Percent Purity 94%
Chemical Name or Material Triethyl 4-phosphonocrotonate, cis + trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.