Learn More
Tricyclohexylphosphonium Tetrafluoroborate, TRC
Product Code. 30457346
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30457346 1 g 1g
1 options
This item is not returnable. View return policy
Product Code. 30457346 Supplier Toronto Research Chemicals Supplier No. TRCT7782901G
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tricyclohexylphosphonium Tetrafluoroborate
CAS 58656-04-5
Molecular Formula C18H33P . BF4.H
Quantity 1 g
Synonym Tri(cyclohexyl)phosphine tetrafluoroborate, PCy3.HBF4
InChI Formula InChI=1S/C18H33P.BF4/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;2-1(3,4)5/h16-18H,1-15H2;/q;-1/p+1
SMILES F[B-](F)(F)F.C1CCC(CC1)[PH+](C2CCCCC2)C3CCCCC3
IUPAC Name tricyclohexylphosphanium;tetrafluoroborate
Molecular Weight (g/mol) 368.2
Formula Weight 368.2427