Learn More
Triclosan-d3, TRC
Product Code. 30452697
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity Unit Size
30452697 1 mg 1mg
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30452697 Supplier Toronto Research Chemicals Supplier No. TRCT7742521MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Triclosan-d3
CAS 1020719-98-5
Purity Grade Notes HPLC
Molecular Formula C12 2H3 H4 Cl3 O2
Quantity 1 mg
Synonym Triclosan D3 (2,4-dichlorophenoxy D3), Phenol','5-chloro-2-(4,6-dichlorophenoxy-2,3,5-d3)-, 5-Chloro-2-(4,6-dichlorophenoxy-2,3,5-d3)-phenol, Triclosan-d3
InChI Formula InChI=1S/C12H7Cl3O2/c13-7-1-3-11(9(15)5-7)17-12-4-2-8(14)6-10(12)16/h1-6,16H/i1D,3D,5D
SMILES [2H]c1c([2H])c(Oc2ccc(Cl)cc2O)c(Cl)c([2H])c1Cl
IUPAC Name 5-chloro-2-(2,4-dichloro-3,5,6-trideuteriophenoxy)phenol
Molecular Weight (g/mol) 292.56
Formula Weight 290.97
Percent Purity >95