missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Triclabenzadol 98 %

Catalog Number p-7112901
Click to view available options
Cantidad:
1 g
5 g
Este artículo no se puede devolver. Vea la política de devoluciones
68786-66-3
C14H9Cl3N2OS
359.66
NQPDXQQQCQDHHW-UHFFFAOYSA-N
triclabendazole, fasinex, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-1h-benzo d imidazole, triclabendazol inn-spanish, triclabendazolum inn-latin, ccris 8988, 5-chloro-6-2,3-dichlorophenoxy-2-methylthio-1h-benzimidazole, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-benzimidazole, 5-chloro-6-2,3-dichlorophenoxy-2-methylthio benzimidazole, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-1h-benzimidazole
50248
6-cloro-5-(2,3-diclorofenoxi)-2-metilsulfanil-1H-bencimidazol
CSC1=NC2=CC(=C(C=C2N1)Cl)OC3=C(C(=CC=C3)Cl)Cl
Este artículo no se puede devolver. Vea la política de devoluciones
68786-66-3
C14H9Cl3N2OS
359.66
NQPDXQQQCQDHHW-UHFFFAOYSA-N
triclabendazole, fasinex, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-1h-benzo d imidazole, triclabendazol inn-spanish, triclabendazolum inn-latin, ccris 8988, 5-chloro-6-2,3-dichlorophenoxy-2-methylthio-1h-benzimidazole, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-benzimidazole, 5-chloro-6-2,3-dichlorophenoxy-2-methylthio benzimidazole, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-1h-benzimidazole
50248
6-cloro-5-(2,3-diclorofenoxi)-2-metilsulfanil-1H-bencimidazol
CSC1=NC2=CC(=C(C=C2N1)Cl)OC3=C(C(=CC=C3)Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Triclabendazole
Punto de fusión 175.0°C to 176.0°C
Intervalo de porcentaje de ensayo 97.5% min. (HPLC)
Envase Frasco de vidrio
Cantidad 1 g
Información de solubilidad Solubility in water: insoluble.
Formula Weight (peso de la fórmula) 359.66
Porcentaje de pureza 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'