missing translation for 'onlineSavingsMsg'
Learn More

Trichloroethylene, Electronic Grade, 99.5+%

Product Code. 17285826
Change view
Click to view available options
Quantity:
1 L
4 x 4 L
Unit Size:
1 L
16 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17285826 4 x 4 L 16 L
17275826 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17285826 Supplier Thermo Scientific Alfa Aesar Supplier No. 41963.M6
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79-01-6 | C2HCl3 | 131.38 g/mol

Trichloroethylene is used as solvent for paints, dyes, waxes, oils, fats and resins, as an additive in polymerization reactions, as a reactant to prepare various halohydrocarbons (1,1,1,2-tetrafluoroethane) and other chemicals, as a coolant for cutting tantalum and acts as good heat transfer medium. It is used in fire extinguisher fluids and in the recovery of oils and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-01-6
Molecular Formula C2HCl3
Molecular Weight (g/mol) 131.38
MDL Number MFCD00000838
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
Synonym trichloroethylene, trichloroethene, ethene, trichloro, trilene, trichlorethylene, ethinyl trichloride, ethylene trichloride, acetylene trichloride, chlorilen, narcogen
PubChem CID 6575
ChEBI CHEBI:16602
IUPAC Name 1,1,2-trichloroethene
SMILES C(=C(Cl)Cl)Cl

Specifications

Melting Point °C to 85°C
Color Colorless
Density 1.46
Boiling Point 87°C
Flash Point None
Ignition Point 410°C (770°F)
Assay Percent Range >99.5%
Odor Chloroform-like
Refractive Index 1.476
Quantity 4 x 4 L
Beilstein 1736782
Solubility Information Miscible with ether,alcohol,chloroformMiscible with ether,ethanol and chloroform. Very slightly miscible with water.
Formula Weight 131.39
Vapor Pressure 77 hPa (58mm Hg) at 20°C
Grade Electronic
Physical Form Liquid
Chemical Name or Material Trichloroethylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.