missing translation for 'onlineSavingsMsg'
Learn More

TRF-2 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NB110-57130V

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214345

  • 623.09€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

TRF-2 Polyclonal specifically detects TRF-2 in Human, Mouse, Rat, Chinese Hamster, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Especificaciones

Especificaciones

TRF-2
Polyclonal
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF 2, TERF2, TRBF 2, TRBF2, TRF 2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2
Rabbit
59.6 kDa
Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Transcription Factors and Regulators
7014
Store at 4°C in the dark.
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
DyLight 405
50 mM sodium borate with 0.05% sodium azide
TERF2
This TRF-2 Antibody was developed against Baculovirus purified TRF2 protein.
0.1 mL
Primary
Human, Mouse, Rat, Chinese Hamster, Primate
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales