missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Treponema pallidum IgG ELISA Kit

For the detection of IgG antibody to Treponema pallidum in Human Serum or Plasma

Marca:  Abnova™ KA0968.1Kit

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 16056445

  • 263.00€ / 1 set


Fecha estimada de envío
08-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

HRP
Colorimetric
Microwell 12 x 8 x 1, 22mL sample diluent, 1.5mL calibrator, 1.5mL positive control, 1.5mL negative control, 12mL Enzyme Conjugate, 12mL TMB chromogenic Substrate, 12mL stop solution, 25mL Washing concentrate (20X)
96 Tests
Plasma, Serum
96 Assays
Treponema pallidum IgG ELISA Kit is used for the detection of IgG antibody to Treponema pallidum in human serum or plasma.
Qualitative
Treponema pallidum IgG ELISA Kit
RUO
Store the kit at 4°C.
10μL
Videos
Documentos