missing translation for 'onlineSavingsMsg'
Learn More

Trappin-2/Elafin/Skalp Antibody (101), Biotin, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Bio-Techne NBP2-90071B

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215856

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Trappin-2/Elafin/Skalp Monoclonal specifically detects Trappin-2/Elafin/Skalp in Human samples. It is validated for ELISA.
Especificaciones

Especificaciones

Trappin-2/Elafin/Skalp
Monoclonal
Biotin
PBS
Rabbit
Protein A purified
RUO
Primary
No cross-reactivity in ELISA with: Human SLPI Human TGM2
Store at 4°C in the dark.
IgG
ELISA
101
ELISA
cementoin, Elastase-specific inhibitor, ESIWAP four-disulfide core domain protein 14, Peptidase inhibitor 3, peptidase inhibitor 3, skin-derived, PI-3, pre-elafin, protease inhibitor 3, skin-derived (SKALP), Protease inhibitor WAP3, SKALPELAFIN, Skin-derived antileukoproteinase, trappin-2, WAP four-disulfide core domain 14, WAP3MGC13613, WFDC14elafin
This antibody was obtained from a rabbit immunized with purified, recombinant Human Trappin-2/Elafin/Skalp (NP_002629.1; Met1-Gln117).
0.1 mL
Cancer, Immunology
5266
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales