missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ TranslationBlocker Human CTLA4 siRNA

Blocks transcription and translation of CTLA4 from Human.

Marca:  enQuireBio™ QX47-10nmol

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16049241

  • 369.00€ / 10 nanomoles


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_005205
siRNA
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month.
Cytotoxic T-Lymphocyte Associated Protein 4, CTLA-3, CD152, Celiac3, Celiac Disease 3, Cytotoxic T-Lymphocyte-Associated Antigen 4
1493
Lyophilized.
10 nmol
Human
CTLA4
CLL Chronic Lymphoid Leukemia Cells.
Videos
Documentos