Description
Employ a novel method for rapid preparation of chemically-competent E. coli cells from overnight bacterial cultures or bacterial colonies with the Thermo Scientific TransformAid™ Bacterial Transformation Kit. The key component of this system is the unique T-solution, which produces competent cells in a few easy steps. The quick and convenient procedure guarantees transformation efficiencies of more than 107 transformants per μg of plasmid DNA, ideal for routine cloning experiments. The option to use bacterial colonies for preparation of competent cells adds flexibility to any cloning experiment.
The TransformAid Bacterial Transformation Kit can be used with most E. coli strains commonly used for cloning: JM107, JM109, XL1-Blue, SURE, TOPP2, W3110, NM527, AD494, CJ236, GM2163, C600, BL21, M15, NM522, HB101, ER2267.
Highlights
- Efficient – more than 107 transformants per μg of plasmid DNA
- Practical – E. coli competent cells can be prepared from an overnight culture or from bacterial colonies
- Fast – less than 1 hour from the overnight bacterial culture to cell plating. Starting from bacterial colonies, the procedure requires only 2.5 hours
- Easy – all procedures are performed on ice on your bench. Brief centrifugations are carried out in regular microcentrifuges
Applications
- Routine cloning experiments
- Blunt-end cloning
- TA cloning
Includes
- C-Medium
- T-Solution (A)
- T-Solution (B)
- Detailed Protocol
Note
Competent cells prepared with TransformAid Bacterial Transformation Kit are suitable for direct-use only. Freezing down and storage at -70°C is not recommended.
Spécification
Spécification
| For Use With (Application) | Routine cloning experiments.Blunt-end cloning.TA cloning. |
| Product Type | TransformAid Bacterial Transformation Kit |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.