Learn More
Transferrin Antibody (HTF-14), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NB500-418UV
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214070
  • 497.83€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
Transferrin Monoclonal specifically detects Transferrin in Human, Porcine, Rabbit, Bovine (Negative), Canine (Negative), Equine (Negative), Sheep (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
Transferrin
Monoclonal
DyLight 350
Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, transferrin
Mouse
Ion exchange chromatography
RUO
Primary
The antibody HTF-14 recognizes an epitope located in the N-terminal domain of human serum transferrin, a 77 kDa single polypeptide chain glycoprotein (member of the iron binding family of proteins). It is synthesised in the liver and consists of two domains each having a high affinity reversible binding site for Fe3+.
Store at 4C in the dark.
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
HTF-14
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
TF
Purified porcine transferrin.
0.1 mL
Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers
7018
Human, Porcine, Rabbit, Bovine (Negative), Canine (Negative), Equine (Negative), Sheep (Negative)
IgG1