Learn More
Ácido trans-4-hidroxi-3-metoxicinámico, 99 %, Thermo Scientific Chemicals
Catalog Number p-7021527
Click to view available options
Cantidad:
25 g
100 g
500 g
537-98-4
C10H10O4
194.19
MFCD00004400
KSEBMYQBYZTDHS-HWKANZROSA-N
ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl
445858
CHEBI:17620
(2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid
COC1=CC(\C=C\C(O)=O)=CC=C1O
537-98-4
C10H10O4
194.19
MFCD00004400
KSEBMYQBYZTDHS-HWKANZROSA-N
ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl
445858
CHEBI:17620
(2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid
COC1=CC(\C=C\C(O)=O)=CC=C1O
A chemopreventive analogue of caffeic acid.trans-4-Hydroxy-3-methoxycinnamic acid is used as an important raw material for the preparation of its corresponding esters by reacting with alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
Análogo quimiopreventivo del ácido cafeico. El ácido trans-4-hidroxi-3-metoxicinámico se utiliza como una materia prima importante para la preparación de sus ésteres correspondientes al reaccionar con alcoholes.
Solubilidad
Soluble en alcohol y agua caliente.
Notas
Incompatible con agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material trans-4-Hydroxy-3-methoxycinnamic acid
Punto de fusión 170°C to 175°C
Cantidad 25 g
Beilstein 1371483
Índice Merck 14,4062
Información de solubilidad Soluble in alcohol and hot water.
Formula Weight (peso de la fórmula) 194.19
Porcentaje de pureza 99%