Learn More
trans-4-Aminocyclohexanol Hydrochloride, Mikromol™
Product Code. 30051752
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30051752 100 mg 100mg
1 options
This item is not returnable. View return policy
Product Code. 30051752 Supplier Mikromol Supplier No. MM0005.06
 Restricted Item
Fecha estimada de envoi: 30-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
trans-4-Aminocyclohexanol Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material trans-4-Aminocyclohexanol Hydrochloride
CAS 50910-54-8
Analyte or Component Names trans-4-Aminocyclohexanol Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C6 H13 N O . Cl H
Certifications/Compliance ISO 9001
Synonym trans-4-Aminocyclohexanol Hydrochloride
InChI Formula InChI=1S/C6H13NO.ClH/c7-5-1-3-6(8)4-2-5;/h5-6,8H,1-4,7H2;1H/t5-,6-;
SMILES Cl.N[C@@H]1CC[C@@H](O)CC1
IUPAC Name 4-aminocyclohexan-1-ol;hydrochloride
Molecular Weight (g/mol) 151.63
Quantity 100 mg
Formula Weight 151.0764 g/mol