missing translation for 'onlineSavingsMsg'
Learn More

Ácido trans-3-(Boc-amino) ciclohexanecarboxílico, 97 %, Thermo Scientific Chemicals

Catalog Number p-7041465
Click to view available options
Cantidad:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
218772-92-0
C12H21NO4
243.30
MFCD03788640,MFCD24395695
JSGHMGKJNZTKGF-UHFFFAOYNA-N
trans-3-tert-butoxycarbonylaminocyclohexanecarboxylic acid, 1r,3r-3-tert-butoxy carbonyl amino cyclohexane-1-carboxylic acid, cyclohexanecarboxylicacid, 3-1,1-dimethylethoxy carbonyl amino-, 1r,3r-rel, trans-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, boc-trans-3-aminocyclohexanecarboxylic acid, trans-3-boc-amino-cyclohexanecarboxylic acid, trans-3-tert-butoxycarbonylaminocyclohexane-carboxylic acid, trans-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3r-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, trans-3-tert-butoxycarbonylamino-cyclohexanecarboxylic acid
24720881
CC(C)(C)OC(=O)NC1CCCC(C1)C(O)=O
Este artículo no se puede devolver. Vea la política de devoluciones
218772-92-0
C12H21NO4
243.30
MFCD03788640,MFCD24395695
JSGHMGKJNZTKGF-UHFFFAOYNA-N
trans-3-tert-butoxycarbonylaminocyclohexanecarboxylic acid, 1r,3r-3-tert-butoxy carbonyl amino cyclohexane-1-carboxylic acid, cyclohexanecarboxylicacid, 3-1,1-dimethylethoxy carbonyl amino-, 1r,3r-rel, trans-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, boc-trans-3-aminocyclohexanecarboxylic acid, trans-3-boc-amino-cyclohexanecarboxylic acid, trans-3-tert-butoxycarbonylaminocyclohexane-carboxylic acid, trans-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,3r-3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, trans-3-tert-butoxycarbonylamino-cyclohexanecarboxylic acid
24720881
CC(C)(C)OC(=O)NC1CCCC(C1)C(O)=O

2-Bromo-4-fluorobenzylamine hydrochloride used as a intermediate in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El clorhidrato de 2-Bromo-4-fluorobenzilamina se utiliza como intermediario en la investigación farmacéutica y química.

Notas
Almacenar en un lugar fresco y bien ventilado. Mantener el recipiente herméticamente cerrado. Almacenar lejos de agentes oxidantes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material trans-3-(Boc-amino)cyclohexanecarboxylic acid
Nombre IUPAC Ácido (1R,3R)-3-[(2-metilpropan-2-il)oxicarbonilamino]ciclohexano-1-carboxílico
Formula Weight (peso de la fórmula) 243.29
Porcentaje de pureza 97%
Cantidad 250 mg

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'