missing translation for 'onlineSavingsMsg'
Learn More

Ácido trans-2,4-difluorocinámico, 97 %

Catalog Number p-7024087
Click to view available options
Cantidad:
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
94977-52-3
C9H6F2O2
184.142
MFCD00010317
PQDXPFJQTKGTFP-DUXPYHPUSA-N
2,4-difluorocinnamic acid, e-3-2,4-difluorophenyl acrylic acid, trans-2,4-difluorocinnamic acid, 3-2,4-difluorophenyl acrylic acid, 2e-3-2,4-difluorophenyl prop-2-enoic acid, 2,4-difluorocinnamicacid, 3-2,4-difluorophenyl-2-propenoic acid, 2e-3-2,4-difluorophenyl acrylic acid, rarechem bk hw 0121, e-3-2,4-difluorophenyl prop-2-enoic acid
5374945
Ácido (E)-3-(2,4-difluorofenil)prop-2-enoico
C1=CC(=C(C=C1F)F)C=CC(=O)O
Este artículo no se puede devolver. Vea la política de devoluciones
94977-52-3
C9H6F2O2
184.142
MFCD00010317
PQDXPFJQTKGTFP-DUXPYHPUSA-N
2,4-difluorocinnamic acid, e-3-2,4-difluorophenyl acrylic acid, trans-2,4-difluorocinnamic acid, 3-2,4-difluorophenyl acrylic acid, 2e-3-2,4-difluorophenyl prop-2-enoic acid, 2,4-difluorocinnamicacid, 3-2,4-difluorophenyl-2-propenoic acid, 2e-3-2,4-difluorophenyl acrylic acid, rarechem bk hw 0121, e-3-2,4-difluorophenyl prop-2-enoic acid
5374945
Ácido (E)-3-(2,4-difluorofenil)prop-2-enoico
C1=CC(=C(C=C1F)F)C=CC(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material trans-2,4-Difluorocinnamic acid
Punto de fusión 215°C to 219°C
Cantidad 5 g
Beilstein 3129944
Formula Weight (peso de la fórmula) 184.14
Porcentaje de pureza 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'