Learn More
TRAIL/TNFSF10 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Marca:  Bio-Techne NB500-220SS
Detalles adicionales : Peso : 0.00970kg
 Ver más versiones de este producto
Código de producto. 18215974
  • 190.06€ / 25 microgramos
Fecha estimada de envío: 14-08-2024
para ver el stock
Descripción
Descripción
TRAIL/TNFSF10 Polyclonal specifically detects TRAIL/TNFSF10 in Human, Mouse samples. It is validated for Western Blot.
Especificaciones
Especificaciones
TRAIL/TNFSF10
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
TNFSF10
residues [SNTLSSPNSKNE] of the N terminus of the TRAIL protein.
Affinity Purified
RUO
Primary
Reacts with residues [SNTLSSPNSKNE] of the N terminus of the TRAIL protein.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot
1 mg/mL
Western Blot 1:100-1:2000
Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10
Rabbit
36 kDa
0.025 mg
Apoptosis, Cancer, Cell Biology, Signal Transduction, Tumor Suppressors
8743
Human, Mouse