missing translation for 'onlineSavingsMsg'
Learn More

TRAcP (Tartrate-Resistant Acid Phosphatase) (Hairy Cell Leukemia Marker) Mouse, Clone: rACP5/1070, enQuireBio™

Mouse Monoclonal Antibody

Marca:  EnquireBio 54-MSM3-P0

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15910658

  • 214.00€ / 20 microgramos


Fecha estimada de envío
16-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

TRAcP (Tartrate-Resistant Acid Phosphatase) (Hairy Cell Leukemia Marker)
Cytoplasmic (Lysosomes)
200μg/mL
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Recombinant full-length human ACP5 protein
35kDa
Protein A/G purified from Bioreactor concentrate.
RUO
Primary
54
Immunohistochemistry
rACP5/1070
Unconjugated
Mouse
IgG1
HepG2, 293T, K562 or RPMI-8226 Cells. Spleen from Hairy Cell Leukemia (HCL) patient.
20 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Videos
Documentos