missing translation for 'onlineSavingsMsg'
Learn More

TRACP/PAP/ACP5 Antibody (rACP5/1070), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-75780IR

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215807

  • 578.82€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

TRACP/PAP/ACP5 Monoclonal specifically detects TRACP/PAP/ACP5 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

TRACP/PAP/ACP5
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
ACP5
Recombinant full-length human TRACP/PAP/ACP5 protein (Uniprot: P13686)
0.1 mL
Protein Phosphatase
It recognizes a protein of 35kDa, which is identified as tartrate-resistant acid phosphatase (TRAcP). It exists as two isoforms (5a and 5b). This monoclonal antibody reacts with both the isoforms. Serum TRAcP 5a is secreted by macrophages and dendritic cells and increased in many patients of rheumatoid arthritis.Serum TRAcP 5b is produced from osteoclasts and elevated during bone resorption. TRAcP is an iron containing glycoprotein, which catalyzes the conversion of orthophosphoric monoester to alcohol and orthophosphate. It is the most basic of the acid phosphatases and is the only form not inhibited by L(+)-tartrate. TRAcP is synthesized as a latent proenzyme and is activated by proteolytic cleavage and reduction. Normally, TRAcP is highly expressed by osteoclasts, activated macrophages, neurons and endometrium during pregnancy. Expression of TRAcP is increased in certain pathological conditions such as Leukemic Reticuloendotheliosis (Hairy Cell Leukemia), Gauchers Disease, HIV-induc
Store at 4°C in the dark.
Immunohistochemistry, Immunohistochemistry (Paraffin)
rACP5/1070
Immunohistochemistry, Immunohistochemistry-Paraffin
HPAP, TRACP5a, TRACP5b, TRAP, TrATPase
Mouse
Protein A or G purified
RUO
Primary
Human, Mouse, Rat
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales