missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TPPP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15057-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody detects the 19-20 kDa TPPP3 in HeLa and PC-3 cells. The non-specific bands between 40 and 60 kDa were also occasionally observed.
TPPP3 binds tubulin and has microtubule bundling activity. TPPP3 may play a role in cell proliferation and mitosis.Especificaciones
TPPP3 | |
Polyclonal | |
Unconjugated | |
TPPP3 | |
CGI 38, p20, p25gamma, TPPP/p20, TPPP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
291966, 51673, 67971 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5PPN5, Q9BW30, Q9CRB6 | |
TPPP3 | |
TPPP3 Fusion Protein Ag7040 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |