Learn More
Lonza™ ToxiLight™ Non-Destructive Cytotoxicity BioAssay Kit, 500 test
Product Code. 11640261
Change view
Click to view available options
Quantity:
5 x 10 mL LT27-060 AK Detection Reagent, 1 x 50 mL LT27-066 AK Assay Buffer
Unit Size:
Each
Product Code. Quantity Unit Size
11640261 5 x 10 mL LT27-060 AK Detection Reagent, 1 x 50 mL LT27-066 AK Assay Buffer Each
1 options
This item is not returnable. View return policy
Product Code. 11640261 Supplier Lonza™ Supplier No. LT07217
  • ONLINE EXCLUSIVE PRICE
    785.00€
    517.84€ / paquete de 1
    Ahorro 267.16€
    34% Descuento
 Restricted Item
Fecha estimada de envoi: 23-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Designed to measure toxicity in mammalian cells and cell lines in culture.
  • Safe - No hazardous chemicals are required.
  • Flexible - Cell supernatants can be frozen with no loss of AK activity, allowing for long-term kinetic studies.
  • Fast - Readable signal is detectable 5 minutes after the addition of the ToxiLight™ BioAssay reagent.
  • Simple - Assay requires the simple addition of a single reagent, which can be added directly to wells in which cells are growing, or to a small sample of aspirated culture supernatant.
  • Non-destructive - ToxiLight™ assay detects cellular AK present in cell culture supernatants, eliminating the need to lyse cells to perform the assay, and allowing multiple tests to be performed on the same sample.
  • Highly sensitive - The ToxiLight™ BioAssay Kit exploits the cyclic nature of the AK reaction whereby a small amount of the AK enzyme can generate high concentrations of ATP. This enables the detection of very subtle changes in cytotoxicity and reduces false negatives.
TRUSTED_SUSTAINABILITY
Specifications
Assays Non-destructive cytotoxicity
Content And Storage LT27-060 AK detection reagent (lyophilized), LT27-066 AK assay buffer
Description 100 tests/ 5 plates
For Use With (Application) Cytotoxicity assay
Quantity 5 x 10 mL LT27-060 AK Detection Reagent, 1 x 50 mL LT27-066 AK Assay Buffer