Learn More
Diversey™ Touchless Dispenser
Product Code. 17613105
Change view
Click to view available options
For Use With (Equipment):
ClearKlens IPA
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17613105 ClearKlens IPA Each
1 options
This item is not returnable. View return policy
Product Code. 17613105 Supplier Diversey™ Supplier No. 4400420
  • ONLINE EXCLUSIVE PRICE
    585.20€
    536.77€ / paquete de 1
    Ahorro 48.43€
    8% Descuento
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Touchless dispenser for use with ClearKlens IPA.
  • Pump technology proven in continuous use
  • Selection of materials according to hygienic viewpoints
  • Touchless operation optimizes hand hygiene
  • Simple pump removal from the front
  • Battery level indicator
  • Suitable for Sterile ClearKlens™ IPA bottles
  • Lockable cover available as an accessory
Body
  • High hygienic standard through anodized aluminum surfaces
  • Stress resistant construction optimized for continuous use
  • Exchangeable face shield for individual product labelling
Pump and dosage
  • IMP Touchless stainless steel pump, angled stainless steel suction tube, autoclavable
  • Dosage amount: approx. 1.5 mL/push, reducible to approx. 0.5 mL/push
Filling: Designed to be used with ClearKlens IPA (70% IPA, 30% WFI)
Specifications
Product Type Touch-free Dispenser
For Use With (Equipment) ClearKlens IPA