missing translation for 'onlineSavingsMsg'
Learn More

Torsin A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10296-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816593

  • 109.00€ / 20 microlitros

Envío estimado: 02-11-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

A mutation of the DYT1 gene, which codes for TorsinA, has been identified as the cause of one form of autosomal domitly inherited dystonia. Early-onset torsion dystonia is a movement disorder, characterized by twisting muscle contractures, that begins in childhood. Symptoms are believed to result from altered neuronal communication in the basal ganglia. TorsinA comprises 332 amino acids. TorsinA is widely expressed throughout the mouse central nervous system and is detected in the majority of neurons in nearly all regions. The proteins display cytoplasmic distribution, although in some types of neurons localization is perinuclear. TorsinA often performs chaperone-like functions that assist in the assembly, operation, or dis-assembly of protein complexes. The gene which encodes TorsinA has high homology to three additional mammalian genes and a nematode gene and distal similarity to the family of heat-shock proteins and the Clp protease family. The gene which encodes TorsinA maps to human chromosome 9q34.
Especificaciones

Especificaciones

Torsin A
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O14656, Q68G38, Q9ER39
DQ2, Dystonia 1 protein, DYT1, TOR1A, Torsin 1A, Torsin A, Torsin family 1 member A, TorsinA
Rabbit
IgG
20 μL
-20°C
Primary
1861, 266606, 30931
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
TOR1A
Liquid
TOR1A
Torsin A Fusion Protein Ag0358
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat, Human, Mouse
Videos
SDS
Documentos
Certificados