missing translation for 'onlineSavingsMsg'
Learn More

Topoisomerase II α Ab-1, Rabbit Polyclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ Topoisomerase II α Ab-1, Rabbit Polyclonal Antibody.

Marca:  Epredia RB-117-P0

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12682807

Cantidad Precio
1 158.00€ / 100 microlitros
10 149.40€ / 100 microlitros
Envío estimado: 01-07-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.



Descripción

Descripción

Topo IIα plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is reported to be a sensitive and specific marker of late S-, G2- & M-phases in transformed and developmentally regulated normal cells. Note that Ki67/M1B1 is also present in G1-phase. Topo IIα is also implicated in drug resistance of tumor cells.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. Others not known.

Epitope: aa 1513-1530

Immunogen: A synthetic peptide, corresponding to aa 1513-1530 (RAKKPIKYLEESDEDDLF) from the C-terminus of human topoisomerase IIα

Molecular Weight: 170kDa

Positive Control: Raji cells or any exponentially growing cultured human cells

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

Topoisomerase II alpha Ab-1
Polyclonal
Unconjugated
Concentrated
A synthetic peptide, corresponding to aa 1513-1530 (RAKKPIKYLEESDEDDLF) from the C-terminus of human topoisomerase IIalpha
100μL
DNA Repair
Primary
Human
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1mg/mL
Purified Ab with BSA and Azide
Rabbit
IgG
RUO
Primary
Polyclonal
Videos
SDS
Documentos
Certificados