missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOP2B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20549-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The antibody is specific to TOP2B.
FOXO4 (Forkhead box O4) is encoded by the FOXO4 gene located on the X chromosome. It is also known as AFX1 or MLLT7 (Myeloid/Lymphoid or Mixed-Lineage Leukemia (Trithorax Homolog, Translocated To, 7) and is a member of the O class of forkhead/ winged helix family of transcription factors along with FOXO1, FOXO3 and FOXO6. FOXO4 is abundantly expressed in skeletal muscle and adipose tissue. FOXO4 binds to the Insulin Responsive Element (IRE) and activates downstream targets of Insulin/IGF1 signaling such as IGFBP1. However, the activation of another downstream target of the Insulin/IGF1 pathway, Akt, leads to phosphorylation of FOXO4 on specific Ser/Thr residues. This phosphorylation mediates binding of FOXO4 to 14-3-3 and eventual exit of FOXO4 from the nucleus.Especificaciones
TOP2B | |
Polyclonal | |
Unconjugated | |
TOP2B | |
DNA topoisomerase 2 beta, TOP2B, top2beta, TOPIIB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21974, 7155 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q02880, Q64511 | |
TOP2B | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |