missing translation for 'onlineSavingsMsg'
Learn More

TOMM20 Antibody (1B2), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP2-60248AF750

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215250

  • 562.62€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

TOMM20 Monoclonal antibody specifically detects TOMM20 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Especificaciones

Especificaciones

TOMM20
Monoclonal
Alexa Fluor 750
KIAA0016Outer mitochondrial membrane receptor Tom20, MAS20, MGC117367, Mitochondrial 20 kDa outer membrane protein, mitochondrial import receptor subunit TOM20 homolog, MOM19, TOM20, translocase of outer mitochondrial membrane 20 homolog (yeast), translocase of outer mitochondrial membrane 20 homolog type II
Recombinant human Tomm20 (25-145aa) purified from E. coli
0.1 mL
Cancer, Endocrinology, Signal Transduction
9804
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, ELISA, Immunofluorescence
1B2
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales