missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMPRSS3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17953-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene encodes a protein that belongs to the serine protease family. The encoded protein contains a serine protease domain, a transmembrane domain, a LDL receptor-like domain, and a scavenger receptor cysteine-rich domain. Serine proteases are known to be involved in a variety of biological processes, whose malfunction often leads to human diseases and disorders. This gene was identified by its association with both congenital and childhood onset autosomal recessive deafness. This gene is expressed in fetal cochlea and many other tissues, and is thought to be involved in the development and maintece of the inner ear or the contents of the perilymph and endolymph. This gene was also identified as a tumor associated gene that is overexpressed in ovarian tumors. Alternatively spliced transcript variants have been described.Especificaciones
TMPRSS3 | |
Polyclonal | |
Unconjugated | |
TMPRSS3 | |
DFNB10, DFNB8, ECHOS1, Serine protease TADG 12, TADG12, TMPRSS3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140765, 309665, 64699 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P57727, Q8K1T0 | |
TMPRSS3 | |
TMPRSS3 Fusion Protein Ag12435 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |