missing translation for 'onlineSavingsMsg'
Learn More

TMEM49 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-88442

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216227

  • 455.71€ / 100 microlitros
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

TMEM49 Polyclonal specifically detects TMEM49 in Mouse samples. It is validated for Western Blot.
Especificaciones

Especificaciones

TMEM49
Polyclonal
Unconjugated
PBS, 2% Sucrose
Rabbit
Affinity purified
RUO
Primary
Mouse
Purified
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
DKFZp566I133, EPG3, TDC1, transmembrane protein 49TMEM49, vacuole membrane protein 1
The immunogen is a synthetic peptide directed towards the C-terminal region of mouse TMEM49
100 μL
Autophagy, Cancer, ER Markers, Membrane Trafficking and Chaperones, Neurodegeneration
81671
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales