Learn More
TMEM49 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca:  Bio-Techne NBP2-78784AF594
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214168
  • 588.54€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
TMEM49 Polyclonal specifically detects TMEM49 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
TMEM49
Polyclonal
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
DKFZp566I133, EPG3, TDC1, transmembrane protein 49TMEM49, vacuole membrane protein 1
Rabbit
Affinity Purified
Autophagy, Cancer, ER Markers, Membrane Trafficking and Chaperones, Neurodegeneration
81671
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 594
50mM Sodium Borate with 0.05% Sodium Azide
VMP1
Synthetic peptide made to an N-terminal portion of human TMEM49 (between amino acids 1-75) [UniProt Q96GC9]
0.1 mL
Primary
Human, Mouse