missing translation for 'onlineSavingsMsg'
Learn More

TMEM214 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Novus Biologicals NBP2-82034AF594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214307

  • 577.74€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

TMEM214 Polyclonal antibody specifically detects TMEM214 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Especificaciones

Especificaciones

TMEM214
Polyclonal
50mM Sodium Borate
Rabbit
Peptide affinity purified
RUO
54867
Store at 4°C in the dark.
IgG
Western Blot, ELISA
Alexa Fluor 594
FLJ20254, FLJ39682, PP446, transmembrane protein 214
TMEM214 antibody was raised against a 17 amino acid synthetic peptide near the center of human TMEM214. The immunogen is located within amino acids 150 - 200 of TMEM214. Amino Acid Squence: HTHDYPYSLVSRELRGI
0.1 mL
Primary
Human, Mouse, Rat
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales