Learn More
TMEM173/STING Rabbit, anti-Human, Unconjugated, Clone: 7K7, Proteintech
Rabbit Recombinant Monoclonal Antibody
Marca: Proteintech 80165-1-RR-20UL
Detalles adicionales : Peso : 0.00020kg
Descripción
TMEM173, also named as ERIS, MITA, STING and MPYS, is a facilitator of innate immune signaling that promotes the production of type I IFN (IFN-alpha and IFN-beta). TMEM173 mediates death signaling via activation of the extracellular signal-regulated kinase (ERK) pathway.
Especificaciones
TMEM173/STING | |
Monoclonal | |
1000 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse, Rat |
Western Blot, Immunohistochemistry, ELISA | |
7K7 | |
Unconjugated | |
20 μL | |
340061 | |
Liquid |