missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM141 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16092-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
TMEM141 is a protein coding gene.Especificaciones
TMEM141 | |
Polyclonal | |
Unconjugated | |
TMEM141 | |
RP11 216L13.7, TMEM141, transmembrane protein 141 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
499755, 51875, 85014 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2AJB2, Q96I45 | |
Tmem141 | |
TMEM141 Fusion Protein Ag8341 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |