missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26877-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The Hox proteins play a role in patterns of embryonic development and cellular differentiation by regulating downstream target genes. TLX1, also called homeobox11 (HOX11), as it is located outside of the four mammalian Hox clusters, is a DNA-binding nuclear transcription factor. TLX1 encodes a homeobox-domain containing protein and is containing a glycine and proline-rich amino terminus. TLX1 is required for maintece of the developing spleen and cell survival. TLX1 is highly expressed in T-cell leukemia and lymphoid neoplasias as a result of translocation. TLX1-deficient mice have no spleen.Especificaciones
TLX1 | |
Polyclonal | |
Unconjugated | |
TLX1 | |
Homeobox protein Hox 11, HOX11, Proto oncogene TCL 3, T cell leukemia homeobox 1, TCL3, TLX1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21908, 3195 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P31314, P43345 | |
TLX1 | |
TLX1 Fusion Protein Ag21899 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |