Learn More
Lovibond™ Tintometer™ Potassium Iodide Tablets
Product Code. 15321985
Change view
Click to view available options
Quantity:
100 Tablets
250 Tablets
Unit Size:
Pack of 100
Pack of 250
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15321985 250 Tablets Pack of 250
15311985 100 Tablets Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15321985 Supplier Lovibond™ Supplier No. 501211
 Restricted Item
Fecha estimada de envoi: 14-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
  • Potassium Iodide Tablets
  • Available in bottles of 100 and 250
Specifications
For Use With (Application) Water Testing
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Quantity 250 Tablets
Type Potassium Iodide Tablets