Learn More
Lovibond™ Tintometer™ PHMB Test Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Marca:  Lovibond™ 515891BT
Detalles adicionales : Peso : 0.07500kg
 Ver más versiones de este producto
Código de producto. 15311895
  • 46.90€ / paquete de 250
Fecha de envío estimada
del proveedor 13-02-2019
Añadir a la cesta
Descripción
Descripción
  • New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
  • With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
  • Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
  • Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
  • ‘BT’ is added to the end of the code to identify the new style of packaging
  • Simple, time-saving and practical
  • There are no safety risks if the tablets are used in line with the instructions supplied
  • Safety data sheets are available for all reagents
Especificación
Especificación
250 Tablets
Test kits, colorimeters and spectrophotometers
Photometry Tablet Reagents
PHMB Test Tablets
Water Testing
Videos