Learn More
Lovibond™ Tintometer™ DPD No. 1 Nessler Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
46.80€ - 102.00€
Especificaciones
Para utilizar con (aplicación) | Reactivos en comprimidos para fotometría |
---|---|
Para utilizar con (equipo) | Kits de pruebas, colorímetros y espectrofotómetros |
Tipo | Comprimidos Nessler DPD n.º 1 |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15301945
|
Lovibond™
511230BT |
100 comprimidos |
46.80€
paquete de 100 |
Fecha estimada de envío: 24-05-2024
Inicie sesión para ver el stock |
|||||
15311945
|
Lovibond™
511231BT |
250 comprimidos |
102.00€
paquete de 250 |
Fecha estimada de envío: 03-06-2024
Inicie sesión para ver el stock |
|||||
Descripción
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
- Available in Packs of 100 and 2550 Tablets
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Especificaciones
Reactivos en comprimidos para fotometría | |
Kits de pruebas, colorímetros y espectrofotómetros | |
Comprimidos Nessler DPD n.º 1 |