Learn More
Alfa Aesar™ Tin(IV) oxide, nanopowder
Marca: Alfa Aesar™ 044606.18
Detalles adicionales : CAS : 18282-10-5 Peso : 0.05000kg
Cantidad. | 50g |
---|
Identificadores de productos químicos
18282-10-5 | |
150.708 | |
XOLBLPGZBRYERU-UHFFFAOYSA-N | |
29011 | |
dioxotin |
O2Sn | |
MFCD00011244 | |
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride | |
CHEBI:52991 | |
O=[Sn]=O |
Especificación
Tin (IV) oxide | |
O2Sn | |
MFCD00011244 | |
tin dioxide, tin iv oxide, stannic oxide, cassiterite, flowers of tin, stannic dioxide, tin oxide sno2, tin peroxide, white tin oxide, stannic anhydride | |
O=[Sn]=O | |
150.708 | |
CHEBI:52991 | |
Nanopowder | |
Odorless | |
1.9968 | |
1630°C |
18282-10-5 | |
SnO2 | |
14,8778 | |
XOLBLPGZBRYERU-UHFFFAOYSA-N | |
dioxotin | |
29011 | |
150.69 | |
6.95g/mL | |
8 to 10nm APS | |
1800°C to 1900°C (sublimation) | |
50g |