missing translation for 'onlineSavingsMsg'
Learn More

TIMP2 Mouse, Clone: 3A4, enQuireBio™

Mouse Monoclonal Antibody

Marca:  EnquireBio 7077-MSM1-P0

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15910978

  • 199.00€ / 20 microgramos


Fecha estimada de envío
13-01-2020
Añadir a la cesta

Especificaciones

Especificaciones

TIMP2
Cytoplasm and Cell Surface
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
TIMP2
A synthetic peptide from the N-terminus of human TIMP2 protein (aa56-66) (DSGNDIYGNPI)
21kDa
Protein A/G Purified
RUO
Primary
7077
Immunohistochemistry
3A4
Unconjugated
NP_003246
Mouse
IgG2a kappa
HeLa cells. Normal Placenta. Breast, Colon, Endometrial, Prostate or Ovarian Carcinoma.
20 μg
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Monoclonal
Videos
Documentos